物質
304 stainless steel needle
nickel plated hub
包装
pack of 12 ea
メーカー/製品名
Cadence Science 4112
L
3/8 in.
ゲージ
33
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
Hypodermic needles are precision made and designed to ensure strict adherence with U.S. Government specification GG-N-196. Points are machine ground to achieve uniform sharpness. Precision grinding, micro-bead jet blasting, and ultrasonic cleaning assure freedom from burrs and debris. All needle cannula are fabricated of type 304 stainless steel tubing which must pass rigorous quality control prior to acceptance and use.
Standard nickle plated Luer hubs comply with Luer taper specifications ISO 594. This assures an interchangeable fit mating with Luer slips and Luer locks. Contoured hub cavity facilitates cleaning.
Standard nickle plated Luer hubs comply with Luer taper specifications ISO 594. This assures an interchangeable fit mating with Luer slips and Luer locks. Contoured hub cavity facilitates cleaning.
最新バージョンのいずれかを選択してください:
Brad Nelms et al.
Science (New York, N.Y.), 364(6435), 52-56 (2019-04-06)
In multicellular organisms, the entry into meiosis is a complex process characterized by increasing meiotic specialization. Using single-cell RNA sequencing, we reconstructed the developmental program into maize male meiosis. A smooth continuum of expression stages before meiosis was followed by
Global Trade Item Number
カタログ番号 | GTIN |
---|---|
CAD4112-12EA | 4061832516769 |