Merck
BR4630240
BRAND® Dispensette® S Organic, Fixed-volume bottle-top dispenser
volume 10 mL, without recirculation valve
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41121500
NACRES:
NB.01
特徴
DE-M marking
without recirculation valve
メーカー/製品名
BRAND 4630240
容量
10 mL
フィッティング
(GL 28/S 28)
GL 32
GL 38
S 40
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
Dispensette S Organic is ideal for dispensing of organic solvents and acids
特徴および利点
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
その他情報
Fixed Volume- For standard applications. Simple calibration adjustment with a supplied tool.
法的情報
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
最新バージョンのいずれかを選択してください:
試験成績書(COA)
文書ライブラリにアクセスする
プロトコル
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.