Merck
BAF249660007
SP Bel-Art® Splash Shield Wall Mounting Fixture
wall bracket, attaches permanently to vertical surface with screws
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41122800
NACRES:
NB.43
包装
pack of 1 ea
メーカー/製品名
Bel-Art F24966-0007
詳細
No matter how your workspace is configured, these Plexiglas® Splash Shields are designed to provide safety plus comfort (not hot and stuffy like personal face shields), flexibility (they’re fully adjustable), and quality in a design that will adapt to your specific needs.
特徴および利点
  • All shields move side-to-side as well as up and down at the touch of a finger
  • Free precious work space and enhance the performance of every user
  • After selecting desired shield, order appropriate mounting fixtures to suit your work area
法的情報
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Plexiglas is a registered trademark of Arkema
最新バージョンのいずれかを選択してください:
試験成績書(COA)