Skip to Content
Merck
All Photos(2)

Key Documents

528122

Sigma-Aldrich

PI 3,4-K inhibitor, PIK-93

The PI 3,4-K inhibitor, PIK-93, also referenced under CAS 593960-11-3, controls the biological activity of PI 3,4-K. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PI 3,4-K inhibitor, PIK-93, N-(5-(4-Chloro-3-(((2-hydroxyethyl)amino)sulfonyl)phenyl)-4-methyl-2-thiazolyl)-acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16ClN3O4S2
CAS Number:
Molecular Weight:
389.88
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 50 mg/mL, pale yellow

shipped in

ambient

storage temp.

2-8°C

SMILES string

ClC1=CC=C(C2=C(C)N=C(NC(C)=O)S2)C=C1S(NCCO)(=O)=O

InChI

1S/C14H16ClN3O4S2/c1-8-13(23-14(17-8)18-9(2)20)10-3-4-11(15)12(7-10)24(21,22)16-5-6-19/h3-4,7,16,19H,5-6H2,1-2H3,(H,17,18,20)

InChI key

JFVNFXCESCXMBC-UHFFFAOYSA-N

General description

A cell-permeable phenylthiazole compound that acts as a potent, reversible and ATP-competitive PI 3,4-K family selective inhibitor (IC50 = 16, 19, 39 and 64 nM for p110γ, PI 4-KIIIβ, p110α and DNA-PK, respectively). Moderately inhibits p110δ, PI 3-KC2β, hsVPS34, ATM, p110β and PI 4-KIIIα (IC50 = 0.120, 0.140, 0.320, 0.490, 0.590 and 1.1 µM, respectively) with minimal inhibition towards a panel of 36-kinases (IC50 >10 µM). Shown to block ceramide transfer protein-mediated ceramide traffic between endoplasmic reticulum and Golgi in transfected COS-7 cells at 250 nM.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Miller, S., et al. 2010. Science327, 1638.
Fan, Q.W., et al. 2006. Cancer Cell9, 341.
Toth, B., et al. 2006. J. Biol. Chem.281, 36369.
Knight, Z.A., et al. 2006. Cell125, 733.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service