Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

182730

Millipore

ASB-C8Ø

A zwitterionic detergent with an aromatic core that is shown to stabilize and solubilize integral membrane proteins by breaking aggregates.

Synonym(s):

ASB-C8Ø, C8Ø, 4- n-Octylbenzoylamido-propyl-dimethylammonio Sulfobetaine, C8Ø, 4-n-Octylbenzoylamido-propyl-dimethylammonio Sulfobetaine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H40N2O4S
CAS Number:
Molecular Weight:
440.64
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.25

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

off-white

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C23H40N2O4S/c1-4-5-6-7-8-9-12-21-13-15-22(16-14-21)23(26)24-17-10-18-25(2,3)19-11-20-30(27,28)29/h13-16H,4-12,17-20H2,1-3H3,(H-,24,26,27,28,29)

InChI key

AQVJFUZGGMXMED-UHFFFAOYSA-N

General description

A zwitterionic detergent (zwittergent®) containing an aromatic core that has been demonstrated to stabilize and solubilize integral membrane proteins by breaking aggregates. Useful for protein analysis and characterization by two-dimensional gel electrophoresis and mass spectrometry techniques. Reported to be superior to CHAPS (Cat. No. 220201) in solubilizing an ion-channel protein and a G-protein coupled receptor (GPCR). In combination with thiourea and urea, also reported to improve the recovery of hydrophobic proteins from Arabidopsis thaliana.
A zwitterionic detergent with an aromatic core that is shown to stabilize and solubilize integral membrane proteins by breaking aggregates. Useful for protein analysis and characterization by two-dimensional (2-D) gel electrophoresis and mass spectrometry techniques. Reported to be superior to CHAPS (Cat. No. 220201) in solubilizing both an anion-channel and a G-protein coupled receptor (GPCR).

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution store in the refrigerator (4°C). Stock solutions are stable for up to 6 months at 4°C.

Other Notes

Henningsen, R., et al. 2002. Proteomics2, 1479.
Molloy, M.P. 2000. Anal. Biochem.280, 1.
Santoni, V., et al. 2000. Electrophoresis21, 3329.
Rabilloud, T., et al. 1999. Electrophoresis20, 3603.
Santoni, V., et al. 1999. Electrophoresis20, 705.
Herbert, B. 1999. Electrophoresis20, 660.
Chevallet, M., et al. 1998. Electrophoresis19, 1901.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Zwittergent is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

182730-VGM:
182730-5GM:
182730-1GM:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service