Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

901537

Sigma-Aldrich

Aluminum Etchant Type F

Synonym(s):

Aluminum etching solution for Al and Al-Si metallization

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352311
NACRES:
NA.23

form

liquid

General description

Aluminum Etchant Type F is a stable preparation used to etch aluminum or aluminum-silicon metallizations on silicon devices and in integrated circuit applications. Aluminum contacts are defined and interconnections are formed. Aluminum Etchant Type F is formulated with unique properties easily overcome many of the difficulties experienced in aluminum etch processes such as residue. The aluminum metallization and etching process using photo-lithographic techniques is basic to the semiconductor and microelectronic technology.
The Aluminum Etchant Type F is highly compatible with many of commercial photoresists and permit delineation into high resolution patterns. Metal line width of 1 mil and separations less than 5 microns are feasible. The high resolution is practical with the Aluminum Etchant Type F because lifting of photoresist patterns does not occur and undercutting is minimized.

Application

  • Aluminum Etchant Type F should not be used or stored in glass or Pyrex containers.
  • Aluminum metallizations up to 25,000 A are vacuum deposited on the silicon slice, coated with a photoresist, and UV exposed using an appropriate photographic mask. The resist is developed to protect the aluminum where interconnections are desired. Then the unprotected areas of the aluminum are removed by etching with the Aluminum Etchant Type F, followed by a water rinse.
  • Etching time is dependent upon the etchant temperature and the aluminum film thickness. When etching thick aluminum films, a higher etch rate is required; thus a higher etchant temperature should be used. Likewise, for thinner aluminum films, slower etch rates are desired and a lower etchant temperature should be chosen.
  • At a specific etchant temperature, the etching time is given by the following formula:
Etching time (second) + Film Thickness (Å)/ Etch Rate (Å/sec)
where Etch Rate at 25°C 30 Å/sec; at 40°C 80 Å/sec.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

901537-500ML:
901537-BULK:
901537-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service