Documenti fondamentali
57080-U
Supelco
Visi-1 Single SPE Tube Processor
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.21
tecniche
solid phase extraction (SPE): suitable
Livello qualitativo
applicazioni
environmental
food and beverages
Descrizione generale
Visi-1 processor provides two rates of flow control
Our Visi-1 Single SPE Tube Processor provides precise flow control through a single 1mL, 3mL, or 6mL SPE tube.
There is no faster, more convenient, or more reliable method for processing one or a few samples.
Simply fill the SPE tube with the appropriate solution, attach it to the Visi-1 processor, and rotate the knurled knob clockwise. The solution will pass through the tube in a slow, uniform rate, consistent with the most reproducible results for SPE. Use the plunger to expel the last drop of solvent from the tip of the tube. Remove the tube from the processor, introduce the next solution, and repeat the process.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Global Trade Item Number
SKUGTIN
57080-U4061832597423