TMO143761
Nunc® MicroWell 96-well Plate with high flange skirt
Nunclon® Delta surface, polystyrene, with lid, clear, case of 50 ea
Sinonimo/i:
multiwell plate, treated multiwell plate, treated plate
About This Item
Codice UNSPSC:
41122102
NACRES:
NB.13
Prodotti consigliati
Materiali
clear
polystyrene
Confezionamento
case of 50 ea
Produttore/marchio commerciale
Nunc 143761
Misura
96 well
Categorie correlate
Descrizione generale
Obtain consistent and reliable results with Thermo Scientific Nunc MicroWell 96-Well Microplates. With multiple surface modifications, these plates are ideal for cell-based assays as well as biochemical assays. The flat bottom shape is suitable for microscopic and optical measurements. Available in either the high flange skirt type or the low flange pinchbar design, the plates are optimized for a range of robotics, automated readers and liquid handling systems.
Note legali
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)