Documenti fondamentali
MTCP4406P
MTC Bio SureStand Multiple Pipette Stand
to hold, 6 x Halo single pipettes OR 6 x ProPette LE single pipettes OR 4 x ProPettemultipipettes
Sinonimo/i:
pipette holder, pipette rack, plastic tube
About This Item
Codice UNSPSC:
41122801
NACRES:
NB.02
Materiali
acrylic
Caratteristiche
to hold 6 x Halo single pipettes OR 6 x ProPette LE single pipettes OR 4 x ProPettemultipipettes
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
MTC Bio P4406-P
Descrizione generale
SureStand Pipette Stand for 6 Halo Single Channel Pipettes or 6 ProPette LE Single Channel Pipettes, up to four ProPette LE Multi-channel Pipettes
SureStand Pipette racks are unique in their ability to accommodate multi-channel pipettes without sacrificing any of the stability of a traditional acrylic rack. The patented “windows” allow the user to insert the multi-channel head longways through the rack while keeping the body of the pipette stable by securing the pipette at two points: under the finger hook and near the bottom of the handle. Each SureStand is also outfitted with four non-skid rubber “feet” for extra stability. SureStand is guaranteed to be the sturdiest and most versatile pipette rack in your lab.
Caratteristiche e vantaggi
  • Universal – Fits all popular pipettes
  • 5mm acrylic – 40% thicker
  • Non-skid rubber feet
  • Multi-channel “window” slots
  • US Patent No 872,300
Note legali
Halo is a trademark of MTC Bio
MTC is a trademark of MTC Bio
ProPette is a trademark of MTC Bio
SureStand is a trademark of MTC Bio
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)