Key Documents
Z201057
Spilfyter® hazmat spill control, polypropylene
Pad, W × L 12 in. × 12 in.
Sinonimo/i:
Hazmat Spill Control
About This Item
Codice UNSPSC:
47131905
NACRES:
NB.35
Confezionamento
pkg of 25 ea
Largh. × lungh.
12 in. × 12 in.
Descrizione generale
Spilfyter® products are made from a specially formulated, lint-free, 100% PP fabric that is virtually inert and will absorb any liquid with which they come in contact. They are able to collect up to 10 times their own weight in waste material (depending upon its viscosity) in 30s. Saturated Spilfyter® products can be landfilled, neutralized, or incinerated in accordance with federal, state and local regulations.
Note legali
Spilfyter is a registered trademark of National Packaging Services Corp.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)