EP0030075455
Eppendorf® BioBased epT.I.P.S.® Reloads
volume 50—1,000 μL, sterile, Biopur, L -71 mm, pkg of 480 ea (5 trays x 96 tips)
About This Item
Prodotti consigliati
Sterilità
sterile
Caratteristiche
DNase free
RNase free
graduations
Biopur
Confezionamento
pkg of 480 ea (5 trays x 96 tips)
Produttore/marchio commerciale
Eppendorf® 0030075455
Caratteristiche più verdi
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
Lunghezza
-71 mm
Volume
50—1,000 μL
Compatibilità
suitable for PCR
suitable for microbiology
suitable for molecular biology
Categoria alternativa più verde
Categorie correlate
Descrizione generale
Applicazioni
Caratteristiche e vantaggi
- Improve your laboratory carbon footprint and switch to epT.I.P.S. BioBased in the new Eppendorf Reload variants. These pipette tips are produced from second-generation biobased polymer with 90% of the feedstock attributed to renewable resources (i.e. waste and residues from vegetable oilrefining or waste cooking oil), avoiding the use of fossil fuel-based petroleum.
- Reduce your laboratory waste volume – use Reloads instead of disposable Racks.
- Eppendorf production facility is ISCC PLUS(International Sustainability & Carbon Certification) certified, confirming the raw materials used at the facility are “bio-circular”. Enables traceabilityalong the entire supply chain.
- Pipette tips with ACT (Accountability, Consistency,and Transparency) Label - an independent environmental impact factor assessmentthrough non-profit My Green.
- Material that is both quality and purity certified.
- Get excellent sample integrity as no slip agents, plasticizers and biocides are used Independent laboratory – Lot-specific purity certificates can be downloaded from Eppendorf website.
- General quality certificates in compliance with standards and general Eppendorf quality items.
- Available in volumes from 10 µL to 1,250 µL Long in Reloads for use with epT.I.P.S. Box 2.0.
- Available as epT.I.P.S. BioBased Biopur, ep Dualfilter T.I.P.S. BioBased PCR clean and sterile, ep Dualfilter T.I.P.S. SealMax BioBased Biopur
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
EP0030075455 | 4065271236684 |