Documenti fondamentali
CLS6796600D
Corning® Digital Hotplates
AC/DC input 230 V AC (Supplied with both 3-pin UK plug and 2-pin Euro plug), plate L × W 10 in. × 10 in., CE compliant
Sinonimo/i:
stirrers
About This Item
Codice UNSPSC:
41102406
NACRES:
NB.72
Caratteristiche
CE compliant
Confezionamento
case of 1
Produttore/marchio commerciale
Corning 6796-600D
Disponibilità
not available in USA and Canada
Parametri
5-550 °C temp. range
Lungh. × largh. lastra
10 in. × 10 in.
Descrizione generale
  • Digital LED temperature display is adjustable in 5 °C increments and blinks until set temperature is reached
  • Microprocessor maintains consistent and repeatable temperature settings.
  • Bright LED HOT TOP icon lights up when top plate temperature is over 60 °C, even when heat control is turned off.
  • Separate temperature sensor provides power cut off if unit overheats
  • Extremely durable and heat resistant Pyroceram glass-ceramic top
  • Small footprint and low profile
  • Meets UL and cUL standards
  • 2 year warranty
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)