Key Documents
CLS352025
Corning® Falcon® Round Bottom Test Tubes
capacity 16 mL, cap, screw, sterile, case of 1000
Sinonimo/i:
culture tube
About This Item
Codice UNSPSC:
41121700
NACRES:
NB.13
Materiali
round polystyrene
Sterilità
sterile
Caratteristiche
cap screw
Confezionamento
case of 1000
Produttore/marchio commerciale
Corning 352025
Capacità
16 mL
Applicazioni
Liquid Handling
Note legali
Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.