XX42PM001
Kit raccordo per manometro
About This Item
Codice UNSPSC:
23151800
eCl@ss:
32031690
Prodotti consigliati
Produttore/marchio commerciale
Pellicon®
Livello qualitativo
Caratteristiche e vantaggi
The Mini holder and Pressure Gauge Adapter Kit is designed for process development and small volume pharmaceutical manufacturing.
Note legali
Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany