Key Documents
Z682675
Ace drying flask, pear-shaped, with side indents
capacity 2000 mL, joint: ST/NS 24/40, glass
Sinonimo/i:
Aldrich® drying flask, pear-shaped, with side indents
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Materiali
glass
pear-shaped flask
single-neck
Giunto
joint: ST/NS24/40
Sterilità
non-sterile
Caratteristiche
closure type Standard Taper Joint
Confezionamento
case of 1
Produttore/marchio commerciale
Büchi equivalent, 11580
Capacità
2000 mL
Descrizione generale
These pear shaped drying flasks for rotary evaporators have side indents which increase the mixing action when the flask is rotated on an evaporator .
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)