Documenti fondamentali
Z562122
Platinum crucible lid
fits 50 mL crucible
Sinonimo/i:
crucibles
About This Item
Codice UNSPSC:
41122200
NACRES:
NB.43
Produttore/marchio commerciale
Birmingham metals
Peso
10.0 g
Descrizione generale
Platinum labware has excellent resistance to acids such as acetic, hydrochloric, hydrofluoric, hydroiodic, nitric, selenic, and sulfuric, and to oxidation at high temperatures. Crucible lids of platinum can be cleaned by boiling in a suitable solvent.
For the removal of substances which have alloyed with the surface, scour with alumina powder.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 10
1 of 10
Platinum crucible lid fits 10 mL crucible
Z562033
Platinum crucible lid
Platinum crucible standard, capacity 25 mL
Z562076
Platinum crucible
Platinum evaporating dish capacity 50 mL
Z562173
Platinum evaporating dish
Platinum crucible standard, capacity 10 mL
Z562025
Platinum crucible
Platinum evaporating dish capacity 100 mL
Z562203
Platinum evaporating dish
Platinum crucible micro, capacity 5 mL
Z562238
Platinum crucible
Global Trade Item Number
SKUGTIN
Z562122-1EA4061833310748