Z557757
Aldrich® Pump N′ Pour can adapter
NPTM 2 in.
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.51
Prodotti consigliati
NPTM
2 in.
Misura
5/8 in. , knurled grip
Descrizione generale
Manufactured of PBT Polybutylene Terephthalate, an extremely tough and chemically resistant plastic material. Use adapter to pour directly from can or to adapt hand pump. See. AL-222 for instructions on installation.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z557757-1EA | 4061826764008 |