Z505943
Nalgene® quick filling/venting closure
number of ports, 3, fits tubing, 1/4 in.
Sinonimo/i:
transfer cap, vented cap
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.35
Prodotti consigliati
Caratteristiche
fits tubing 1/4 in.
number of ports 3
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Nalgene 2158-0031
Descrizione generale
Available in two and three port configurations, and is ideal for filling/dispensing into and out of vacuum trap systems with carboys and bottles. Quick disconnect ports fit either 1/4 in. or 3/8 in. I.D. tubing, and have a built-in shut off valve if tubing inserts are taken out. Fits onto all carboys and bottles with 83 mm neck. Leakproof.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Accessorio
N° Catalogo
Descrizione
Determinazione del prezzo
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)