Z500011
Nalgene® quick filling/venting closure
fits tubing, 3/8 in., number of ports, 3
Sinonimo/i:
transfer cap, vented cap
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.35
Prodotti consigliati
Caratteristiche
fits tubing 3/8 in.
number of ports 3
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Nalgene 2158-0032
Categorie correlate
Descrizione generale
Available in two and three port configurations, and is ideal for filling/dispensing into and out of vacuum trap systems with carboys and bottles. Quick disconnect ports fit either 1/4 in. or 3/8 in. I.D. tubing, and have a built-in shut off valve if tubing inserts are taken out. Fits onto all carboys and bottles with 83 mm neck. Leakproof.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Accessorio
N° Catalogo
Descrizione
Determinazione del prezzo
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z500011-1EA | 4061832618104 |