Z414131
Aldrich® dual bank manifold with high-vacuum PTFE valves
standard manifold, positions, 3
About This Item
Codice UNSPSC:
40161501
NACRES:
NB.79
Prodotti consigliati
Caratteristiche
positions 3
Lunghezza
300 mm
Categorie correlate
Descrizione generale
The dual bank manifolds come with 0-10 mm bore high-vacuum PTFE valves.
Attention: We supply high quality PTFE valves on Aldrich brand glassware; these have been tested and approved by our Glass Center.
Attention: We supply high quality PTFE valves on Aldrich brand glassware; these have been tested and approved by our Glass Center.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Prodotti correlati
N° Catalogo
Descrizione
Determinazione del prezzo
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z414131-1EA | 4061837619861 |