Z219738
Aldrich® manifold with trap
male joint: ST/NS 14/20 (1), male Joints: ST/NS 24/40 (2), PTFE valves
About This Item
Codice UNSPSC:
40161501
NACRES:
NB.79
Prodotti consigliati
Giunto
male Joints: ST/NS24/40 (2)
male joint: ST/NS14/20 (1)
Caratteristiche
1 hose barb connection
Categorie correlate
Descrizione generale
Features four 0-10 mm bore high-vacuum PTFE valves and a removable end cap for easy cleaning. Approx. 580 mm L. In-line trap eliminates extra hookup, thus improving vacuum performance.
NOTICE: We supply high quality PTFE valves on Aldrich brand glassware that has been tested and approved by our Glass Center.
NOTICE: We supply high quality PTFE valves on Aldrich brand glassware that has been tested and approved by our Glass Center.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Prodotti correlati
N° Catalogo
Descrizione
Determinazione del prezzo
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z219738-1EA | 4061832131771 |