Documenti fondamentali
Z290882
Sigma-Aldrich
PEEK Tubing Cutter
Sinonimo/i:
PEEK Tubing Cutter
About This Item
Codice UNSPSC:
41115700
eCl@ss:
32035301
NACRES:
SB.52
Descrizione generale
Makes burr-free, perpendicular cuts through polymer tubing with outside diameters from 1/16 in. to 1/8 in.. Designed for cutting PEEK tubing, but also easily slices through PTFE and Tefzel capillary tubing. Compact design includes a safety locking mechanism. One spare blade included.
Prodotti consigliati
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 4
1 of 4
Straightening Pliers
58646
Straightening Pliers
Upchurch Fittings Kit for use with Waters
58639
Upchurch Fittings Kit
vibrant-m
54994
PEEK Tubing
Global Trade Item Number
SKUGTIN
Z290882-1EA4061838553935