Documenti fondamentali
Z286419
FEP sample tube liner
Wilmad 6010-7, O.D. 10 mm
Sinonimo/i:
Wilmad nmr tubes, nmr sample tubes, nmr tubes
About This Item
Codice UNSPSC:
41121705
NACRES:
SB.32
Descrizione
Recommended outer tube 513-7PP-7
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Wilmad 6010-7
D.E.
10 mm
Volume
440 μL
Descrizione generale
Thin-wall FEP liner minimizes filling factor losses of using inserts and protects NMR tube from etching effects of strong acids containing fluoride ions. Liners are 8 in. L and fit snugly into most common 7 in. L thin-wall tubes. A PTFE plug to close each liner is included. Not recommended for elevated temperature studies.
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)