Documenti fondamentali
Z285889
Sigma-Aldrich
E-Z Press
About This Item
Codice UNSPSC:
12000000
Lungh. × prof. × alt.
7 1/2 in. × 6 in. × 15 1/2 in.
Diam. piatto
4 in.
Peso
49.9 lb
Descrizione generale
• Built-in safety shield
• Ergonomic design
• Light weight
12 ton laboratory press with unibody construction designed to work with the Aldrich macro-micro and similar KBr pellet dies. Built-in pressure gauge and pressure release valve provide accurate control of pressure. Press includes removable handle and base.
Note legali
E-Z Press is a trademark of International Crystal Laboratories Corp.
Prodotti correlati
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)