Documenti fondamentali
Z281379
Laboratory brushes
Burette, L 36 in., brush diam. × L 3/4 in. × 3 1/4 in.
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Confezionamento
pkg of 4 ea
Lunghezza
36 in.
brush diam. × L
3/4 in. × 3 1/4 in.
Descrizione generale
Brush materials resistant to acids, aromatic and aliphatic hydrocarbons, ketones, ethyl acetate esters, trichloroethylene, and hot water. FDA and USDA approved materials.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 4
1 of 4
Laboratory brushes Centrifuge, L 12 in., brush diam. × L 1 1/2 in. × 4 1/2 in.
Z281352
Laboratory brushes
Laboratory brushes Micro tube, L 7 in., brush diam. × L 3/16- 1/2 in. × 3 in.
Z281247
Laboratory brushes
Laboratory brushes Flask, L 12 in., brush diam. × L 1 3/8 in. × 3 1/2 in.
Z281344
Laboratory brushes
Global Trade Item Number
SKUGTIN
Z281379-1PAK4061833352601