Documenti fondamentali
Z268704
Scienceware® Safe-Lab PTFE stopper
joint: ST/NS 24/40, solid
Sinonimo/i:
lab stopper, lab stoppers, stoppers
About This Item
Codice UNSPSC:
41122500
NACRES:
NB.35
Giunto
joint: ST/NS24/40
Confezionamento
pkg of 2 ea
Produttore/marchio commerciale
Bel-Art F20196-1502
Descrizione generale
PTFE stoppers with PP extracting nut for easy removal from flask. For use with boiling and distilling flasks with ground joints.
Note legali
Safe-Lab is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)