Documenti fondamentali
Z232378
Pouring rings for Duran® laboratory bottles
GL 32 neck thread, blue polypropylene
Sinonimo/i:
pouring ring
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Materiali
blue polypropylene
Sterilità
non-sterile
Produttore/marchio commerciale
DURAN®
Schott
Parametri
-40-140 °C temp. stability
Raccordi
GL 32 neck thread
Descrizione generale
The classic laboratory bottle GL threaded screw cap features the tried and tested cap plug sealing system ensuring a liquid tight seal. Available in four colours; blue, green, yellow and grey with matching pouring rings. Distinguishing media types is simplified and interchanging of screw caps and the risk of cross contamination is greatly reduced.
Caratteristiche e vantaggi
Optional pouring rings prevent drips when pouring from Duran bottles.
Note legali
DURAN is a registered trademark of DWK Life Sciences
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Global Trade Item Number
SKUGTIN
Z232378-10EA4061832798608