Documenti fondamentali
Z130974
Nalgene® Griffin beakers
capacity 250 mL, translucent polypropylene
Sinonimo/i:
PP beaker, nalgene PP beaker
About This Item
Codice UNSPSC:
41121803
NACRES:
NB.43
Materiali
translucent polypropylene
Caratteristiche
graduations
Confezionamento
pkg of 6 ea
Produttore/marchio commerciale
Nalgene 1201-0250
Capacità
250 mL
Descrizione generale
Low form. With silk-screened graduations, Nalgene catalog number, size and resin codes, max. use temp., and "no flame" symbol. Meet the ISO/DIS 7056 International Standards for Laboratory Plasticware Accuracy. Chemically resistant. Autoclavable. Not for use on a hot plate.
Altre note
Browse all plastic beakers
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)