Documenti fondamentali
Z126306
Supelco
Bottle-type sprayer
capacity 240 mL
About This Item
Codice UNSPSC:
41115711
NACRES:
NC.07
tecniche
thin layer chromatography (TLC): suitable
Capacità
240 mL
Applicazioni
Bottle-type sprayer may be used in thin-layer chromatography.
Prodotti correlati
N° Catalogo
Descrizione
Determinazione del prezzo
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Fatemeh S Mirnaghi et al.
Analytical chemistry, 83(15), 6018-6025 (2011-06-30)
Biocompatible C18-polyacrylonitrile (PAN) coating was used as the extraction phase for an automated 96-blade solid phase microextraction (SPME) system with thin-film geometry. Three different methods of coating preparation (dipping, brush painting, and spraying) were evaluated; the spraying method was optimum
Articoli
Separation of AMP (adenosine monophosphate); ATP (adenosine triphosphate)
Protocolli
Separation of Fumaric acid; Oxalic acid; Citric acid
Separation of Placebo; 1 % Vit C derivat; 1 % Active 2; 2 % Vit C Derivat; 2 % Active2; Reference Vitamin C Derivat; Reference Degradation product