Documenti fondamentali
Z104361
Tubing quick-disconnects
for tubing i.d., 3/8 ‑ 1/2in.
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
Materiali
PE
Caratteristiche
for tubing i.d. 3/8 ‑ 1/2in.
Confezionamento
pack of 12 ea
Produttore/marchio commerciale
Bel-Art F19729-0000
Categorie correlate
Descrizione generale
Consists of two serrated connectors which are joined securely through a male-female center taper. Connector of one size can be interchanged with other sizes to produce reducing combinations.
  • Easy-to-assemble barbed connectors require no tools
  • Ideal for use with flexible tubing
  • Handy on lines that require frequent disconnection
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Global Trade Item Number
SKUGTIN
Z104361-12EA4061832261409