WHA2017009
Whatman® Antibiotic Assay Discs
diam. 9 mm
Sinonimo/i:
Z741329
About This Item
Codice UNSPSC:
41111700
NACRES:
NB.79
Prodotti consigliati
Confezionamento
pkg of 1000 ea
Produttore/marchio commerciale
Whatman 2017-009
Whatman Article No. 28418661 (US reference)
Diametro
9 mm
Categorie correlate
Descrizione generale
For determining the type of causal agent of infectious diseases and for checking their sensitivity to antibiotics and chemotherapeutic agents in vitro by means of the inhibition zone determination method.
Caratteristiche e vantaggi
- The antibiogram allows rational and selective chemotherapy.
- Test discs can be coated with chemotherapeutic agents, placed on the innoculated nutrient agar and incubated.
- The size of the inhibition zone is a measure for the effectiveness of the substances.
Altre note
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Note legali
Whatman is a registered trademark of Cytiva
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)