Documenti fondamentali
WHA10548232
Whatman® protein saver card accessories
Plastic ziploc bags 4 in. x 6 in., pack of 100 ea
Sinonimo/i:
Z699551, ziploc bag, ziploc bags
About This Item
Codice UNSPSC:
41111700
NACRES:
NB.79
Confezionamento
pack of 100 ea
Produttore/marchio commerciale
Whatman 10548232
Whatman Article No. 28417398 (US reference)
Descrizione generale
These durable bags provide convenient storage and protection for protein saver cards, ensuring safe handling and transport in laboratory environments.
Caratteristiche e vantaggi
  • 4" x 6"
  • 100EA
Altre note
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Note legali
Whatman is a registered trademark of Cytiva
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)