WHA10534321
Whatman® protein saver card accessories
Foil barrier reusable bags, pack of 100 ea
Sinonimo/i:
Z699527, foil bag, foil bags
About This Item
Codice UNSPSC:
41111700
NACRES:
NB.79
Prodotti consigliati
Confezionamento
pack of 100 ea
Produttore/marchio commerciale
Whatman 10534321
Whatman Article No. 28416623 (US reference)
Descrizione generale
These bags effectively safeguard contents from moisture and contaminants while ensuring easy access.
Caratteristiche e vantaggi
- Foil bag with size 3.875” × 5.6875” and zipper to close.
- Suitable for use with Whatman 903 cards.
Altre note
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Note legali
Whatman is a registered trademark of Cytiva
Prodotti correlati
N° Catalogo
Descrizione
Determinazione del prezzo
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)