Documenti fondamentali
TEM-LC200AU25
Sigma-Aldrich
Lacey Carbon Supported Gold Grids
size 200 mesh, size 50 nm , hole size, box of 25 ×
Sinonimo/i:
Lacey Carbon Film - Au, 200 mesh, 25pk
About This Item
Codice UNSPSC:
41111761
NACRES:
MA.24
Confezionamento
box of 25 ×
Misura
200 mesh
50 nm , hole size
Descrizione generale
Lacey carbon film supported TEM grids. This carbon coating consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)