Documenti fondamentali
SYNF544500
Synthware reaction flask with high vacuum valve
500 mL, joint: ST/NS 24/40, valve size 0-4 mm
Sinonimo/i:
Synthware F544500, Schlenk flask
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.51
Materiali
borosilicate glass flask
round bottom
single-neck (high vaccum valve)
Giunto
joint: ST/NS24/40
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware F544500
Capacità
500 mL
500 mL
Misura valvola
0-4 mm
Descrizione generale
Supplied with high vacuum valve with PTFE O-ring at tip and PTFE wiper to protect Viton® O-ring on shaft.
Note legali
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Global Trade Item Number
SKUGTIN
SYNF544500-1EA4061832195070