Documenti fondamentali
SYNF128000
Synthware single-neck flat-bottom flask
capacity 1,000 mL, joint: ST/NS 29/42
Sinonimo/i:
Synthware F128000, glass round flask
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.51
Materiali
borosilicate glass flask
flat bottom
single-neck
Giunto
joint: ST/NS29/42
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware F128000
Capacità
1,000 mL
Descrizione generale
Heavy wall single neck, flat bottom flask with an outer joint. Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)