SLW5809/3
Pyrex® Filter holder for 47 mm membranes
base only
Sinonimo/i:
Pyrex, SciLabware 5809/3
About This Item
Codice UNSPSC:
41123300
NACRES:
NB.43
Prodotti consigliati
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
5809/3
SciLabware
Descrizione generale
Comprising of a 300 ml capacity funnel, support base and metal clamp, this popular size filter holder is designed to be used with 47 mm membrane filters. The three components of this product must be ordered separately.
Note legali
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
SLW5809/3-1EA | 4061832443140 |