SLW1000/06D
Pyrex® Griffin beakers, low form, with printed trace code
150 mL
Sinonimo/i:
Pyrex, SciLabware 1000/06D
About This Item
Codice UNSPSC:
41121803
NACRES:
NB.43
Prodotti consigliati
Materiali
borosilicate glass
white graduation
Caratteristiche
low form
with printed trace code
Produttore/marchio commerciale
1000/06D
SciLabware
Alt. × Diam.
80 mm × 60 mm
Capacità
150 mL
Descrizione generale
- Complies with ISO 3819 and DIN 12331
- Manufactured from Pyrex borosilicate glass
- Excellent thermal performance
- High resistance to chemical attack
- Printed with Trace Code for downloadable batch certificate
- Batch certificate available to download from www.scilabware.com
- White graduations and marking spot
Altre note
Browse all glass beakers
Note legali
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
SLW1000/06D-10EA | 4061832435442 |