Documenti fondamentali
SLW1000/02M
Pyrex® Griffin beakers, low form, with printed trace code
50 mL
Sinonimo/i:
Pyrex, SciLabware 1000/02M
About This Item
Codice UNSPSC:
41121803
NACRES:
NB.43
Materiali
borosilicate glass
white graduation
Caratteristiche
low form
with printed trace code
Produttore/marchio commerciale
SciLabware 1000/02M
Alt. × Diam.
60 mm × 42 mm
Capacità
50 mL
Descrizione generale
  • Complies with ISO 3819 and DIN 12331
  • Manufactured from Pyrex borosilicate glass
  • Excellent thermal performance
  • High resistance to chemical attack
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com
  • White graduations and marking spot
Altre note
Browse all glass beakers
Note legali
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)