Documenti fondamentali
DWK213911772
DURAN® measuring cylinder ungraduated with hexagonal base
glass, cylinder capacity 50 nL, unstamped
Sinonimo/i:
213911772
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Materiali
glass
Caratteristiche
lid: no
Confezionamento
case of 10 ea
Produttore/marchio commerciale
DURAN® 213911772
Capacità del cilindro
50 nL
Descrizione generale
Calibration is based on the poured in volume (“In”) at a + 20 °C reference temperature. A uniform wall thickness over the entire measuring range prevents wedge errors. Fired-on, highly durable white print. Easy-to-read scale with graduation marks to estimate the content. The volume content tolerances conform to accuracy Class B.
Note legali
DURAN is a registered trademark of DWK Life Sciences
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)