Documenti fondamentali
CLS999813
Corning® phenolic cap, PTFE liner
13-415 thread
Sinonimo/i:
Corning cellstack cell culture chambers, black phenolic, phenolic caps, screw caps, tissue culture chambers, Corning® 9998
About This Item
Codice UNSPSC:
41122500
NACRES:
NB.43
Materiali
PTFE liner
black phenolic cap
Produttore/marchio commerciale
Corning 9998-13
Altezza
~13.5 mm
Raccordi
13-415 thread
Descrizione generale
These reusable Corning caps are fabricated from specially formulated resin to resist the effects of temperature and steam in autoclaving. Caps are provided with a glued-in rubber liner over which is firmly bonded a PTFE interface to provide a highly inert sealing face.
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)