Key Documents
CLS9999241
Corning® phenolic cap with white rubber liner
24-410 thread
Sinonimo/i:
phenolic caps, screw caps
About This Item
Codice UNSPSC:
41122500
NACRES:
NB.43
Produttore/marchio commerciale
Corning 9999-241
Raccordi
24-410 thread
Descrizione generale
These reusable Corning caps are fabricated from specially formulated phenolic resin to resist the effects of temperature and steam in autoclaving. Caps are provided with a glued-in white rubber liner.
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)