Documenti fondamentali
CLS814040
Pyrex® heavy-duty graduated centrifuge tube, beaded rim
capacity 40 mL
Sinonimo/i:
Corning® 8140
About This Item
Codice UNSPSC:
41121703
NACRES:
NB.43
Materiali
conical bottom tube
Confezionamento
case of 12
pack of 6
Produttore/marchio commerciale
Corning 8140-40
D.E. × lungh.
28 mm × 135 mm
Capacità
40 mL
Descrizione generale
The 12 mL size is graduated upward from thip in 0.1 mL divisions. The 40 mL size is graduated in 0.5 mL divisions to 10 mL, and from 10 to 40 mL in 1 mL divisions.
Note legali
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)