CAD6186
Adapter, FLL to Pomeroy ear tip (plated brass)
FLL to pomeroy ear tip (plated brass)
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.03
Prodotti consigliati
Confezionamento
pack of 10 ea
Produttore/marchio commerciale
Cadence Science 6186
Descrizione generale
- Plated brass or 316 stainless steel
- Meets ISO 594 specifications
- Final assembly water tested at 45 PSI
Female Luer Lock (FLL) - contains a tabbed hub for secure fitting to a threaded male Luer lock.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
CAD6186-10EA | 4061832517544 |