BR719505
BRAND® counting chamber BLAUBRAND® Bürker-Türk
without clips, double ruled
Sinonimo/i:
BLAUBRAND®, Bürker-Türk, Cell culture, Counting Chambers
About This Item
Codice UNSPSC:
41122600
NACRES:
NB.22
Prodotti consigliati
Materiali
colorless
glass
Sterilità
non-sterile
Caratteristiche
without clips
Produttore/marchio commerciale
BRAND 719505
Pozzetti
1
Descrizione generale
Manufactured according to DIN 12 847, including 2 haemacytometer cover glasses.Combination of the Bürker and Thoma systems.The ruling shows 9 large squares of 1 mm² each. The large squares are subdivided into 16 group squares with 0.2 mm sides. in the central large square, each group square is subdivided into 16 mini squares with 0.05 mm sides (= 0.0025 mm²).
Caratteristiche e vantaggi
- BLAUBRAND® quality
- Outstanding measurement precision
- 100% certified quality
Confezionamento
1 counting chamber, complete with 2 haemacytometer cover glasses, in transparent plastic box.
Note legali
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Rachele Isticato et al.
Environmental microbiology, 22(1), 170-182 (2019-11-13)
Bacterial spores are commonly isolated from a variety of different environments, including extreme habitats. Although it is well established that such ubiquitous distribution reflects the spore resistance properties, it is not clear whether the growing conditions affect the spore structure
Mariamichela Lanzilli et al.
New biotechnology, 46, 38-44 (2018-07-01)
Bacterial spores displaying heterologous proteins have been proposed as a safe and efficient method for delivery of antigens and enzymes to animal mucosal surfaces. Initial studies have been performed using Bacillus subtilis spores, but other spore forming organisms have also