Key Documents
BR718005
BRAND® counting chamber BLAUBRAND® Thoma pattern
without clips, double ruled
Sinonimo/i:
BLAUBRAND®, Cell culture, Counting Chambers, Thoma
About This Item
Codice UNSPSC:
41122600
NACRES:
NB.22
Materiali
colorless
glass
Sterilità
non-sterile
Caratteristiche
without clips
Produttore/marchio commerciale
BRAND 718005
Pozzetti
1
Descrizione generale
Manufactured according to DIN 12 847, including 2 haemacytometer cover glasses.Rulings correspond to the central large square of the Neubauer chamber. The mini squares have an area of 0.0025 mm² each. Since the outer large squares are not completed, the Thoma chamber is only used for counting thrombocytes and erythrocytes.
Caratteristiche e vantaggi
• BLAUBRAND® quality• Outstanding measurement precision• 100% certified quality
Confezionamento
1 counting chamber, complete with 2 haemacytometer cover glasses, in transparent plastic box.
Note legali
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)