Documenti fondamentali
BR717805
BRAND® counting chamber BLAUBRAND® Neubauer improved
without clips, double ruled
Sinonimo/i:
BLAUBRAND®, Cell culture, Counting Chambers, Neubauer, ldpe bottle, polyethylene bottles, polyethylene containers
About This Item
Codice UNSPSC:
41122600
NACRES:
NB.43
Materiali
colorless
glass
Sterilità
non-sterile
Caratteristiche
without clips
Produttore/marchio commerciale
BRAND 717805
Dimens. camera
0.1 mm , depth
Pozzetti
1
Descrizione generale
Manufactured according to DIN 12 847, including 2 haemacytometer cover glasses.The ruling shows 9 large squares of 1 mm² each.The four large squares in the corners marked "L" for counting leucocytes are each subdivided into 16 squares with 0.25 mm sides.The large square in the center is subdivided into 16 squares with 0.25 mm sides.The large square in the center is subdivided into 25 group squares of 0.2 mm sides. Each group square consists of 16 mini squares with 0.05 mm sides, each having an area of 0.0025 mm². The 5 group squares in the large central square are used for counting thrombocytes and erythrocytes.All group squares have triple boundary lines on each side. The central line is the limiting line and determines whether cells in the marginal area shall be included in the count or not.
Caratteristiche e vantaggi
  • BLAUBRAND® counting chambers are precision measuring instruments, used to determine the number of particles per volume unit of a suspension.
  • Same ruling as improved Neubauer, but with rhodium coated chamber bottom.
  • Rulings are engraved into the rhodium layer and appear bright under normal microscope settings.
  • By altering the contrast, the microscope image can be reversed, so that the rulings appear brighter or darker, as required.
Confezionamento
Include 2 hemocytometer cover glasses. The dimensions are 24mm L x 26mm W x 0.4mm H. They are also sold separately as item BR723014.
Note legali
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Manoj Kumar Kanakasabapathy et al.
Lab on a chip, 17(17), 2910-2919 (2017-07-14)
The most recent guidelines have called for a significant shift towards viral load testing for HIV/AIDS management in developing countries; however point-of-care (POC) CD4 testing still remains an important component of disease staging in multiple developing countries. Advancements in micro/nanotechnologies
Martin W Breuss et al.
Nature medicine, 26(1), 143-150 (2019-12-25)
De novo mutations arising on the paternal chromosome make the largest known contribution to autism risk, and correlate with paternal age at the time of conception. The recurrence risk for autism spectrum disorders is substantial, leading many families to decline