BR137750
BRAND® magnetic stirring bar retriever, PP
L 300 mm, diam. 10 mm, with ring
Sinonimo/i:
Magnetic stirring bar retriever, Magnetic stirring bars
About This Item
Codice UNSPSC:
41122402
NACRES:
NB.72
Prodotti consigliati
Caratteristiche
with ring
Produttore/marchio commerciale
BRAND 137750
Lunghezza
300 mm
Diametro
10 mm
Categorie correlate
Descrizione generale
The BRAND® Magnetic Stirring Bar Retriever is an essential tool for efficiently retrieving magnetic stirring bars from containers and vessels. Constructed from durable polypropylene (PP), this retriever features a fully sealed magnetic core.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
BR137750-1EA | 4061831853100 |