BMSBSH5002E
myBlock ll digital dual chamber dry bath without blocks
AC/DC input 230 V AC, Schuko plug
About This Item
Codice UNSPSC:
41102404
NACRES:
NB.76
Prodotti consigliati
Produttore/marchio commerciale
Benchmark Scientific BSH5002-E
Parametri
5-105 °C temp. range
Capacità
, holds 2 blocks
Descrizione generale
Benchmark′s myBlock series is a "next generation" product line that introduces several major advantages over traditional dry baths. They are the first digital dry baths to include advanced microprocessor controls, timed or continuous operation and a removable hinged lid. In addition, an optional external temperature feedback probe can be positioned directly inside a sample tube, providing real-time control and monitoring of actual sample temperature.
- Digital temperature control (set and walk away)
- Hinged lid (improves uniformity & conserves energy)
- External (optional) temperature probe (measures actual sample temperature)
- Available with Quick-Flip universal blocks (supports all common tube sizes, PCR strips and PCR plates)
Prodotti correlati
N° Catalogo
Descrizione
Determinazione del prezzo
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BMSBSH5002E-1EA | 4061832874296 |