主要文件
55085-U
Upchurch 串联单向阀
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41115700
NACRES:
SB.52
相关类别
一般說明
The inline Cartridge Check Valve is designed to limit flow to one direction. The cartridge and flangeless fitting is for use with 1/16″ O.D. tubing. The internal volume of the check valve is less than 150μL. Place in the flow system where you wish to restrict fluid flow to one direction. These assemblies withstand system pressures of 1.000 psi (69 bar). Complete with wrench.
推薦產品
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
历史批次信息供参考:
分析证书(COA)